Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Serine, FCC, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
302-84-1
C3H7NO3
105.09
MTCFGRXMJLQNBG-UHFFFAOYNA-N
2-amino-3-hydroxypropanoic acid
NC(CO)C(O)=O
302-84-1
C3H7NO3
105.09
MTCFGRXMJLQNBG-UHFFFAOYNA-N
2-amino-3-hydroxypropanoic acid
NC(CO)C(O)=O

S1124, 302-84-1, C3H7NO3

Spectrum™ DL-Serine, FCC is used as an amino acid nutritional supplement and flavor enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.003
Quantity 100 g
CAS Max % 1
Percent Purity 98.5 to 101.5%