Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DL-Tartaric acid anhydrous, ≥99.5% (T), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
5 g
Specifications
Melting Point | 210°C to 212°C |
CAS | 133-37-9 |
Linear Formula | HOOC(CHOH)2 COOH |
Solubility Information | Water: 0.1 g/mL, clear |
Formula Weight | 150.09 |
Percent Purity | ≥99.5% (T) |
Grade | Ion Chromatography |
Quantity | 5 g |
Flash Point | 210°C - Closed Cup |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label