Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Tropic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
552-63-6
C9H10O3
166.18
MFCD00004255
JACRWUWPXAESPB-UHFFFAOYNA-N
tropic acid,dl-tropic acid,2-phenylhydracrylic acid,3-hydroxy-2-phenylpropionic acid,+--tropic acid,tropate,alpha-hydroxymethyl benzeneacetic acid,beta-hydroxyhydratropic acid,hydracrylic acid, 2-phenyl,alpha-hydroxymethyl phenylacetic acid
10726
CHEBI:30765
3-hydroxy-2-phenylpropanoic acid
OCC(C(O)=O)C1=CC=CC=C1
552-63-6
C9H10O3
166.18
MFCD00004255
JACRWUWPXAESPB-UHFFFAOYNA-N
tropic acid,dl-tropic acid,2-phenylhydracrylic acid,3-hydroxy-2-phenylpropionic acid,+--tropic acid,tropate,alpha-hydroxymethyl benzeneacetic acid,beta-hydroxyhydratropic acid,hydracrylic acid, 2-phenyl,alpha-hydroxymethyl phenylacetic acid
10726
CHEBI:30765
3-hydroxy-2-phenylpropanoic acid
OCC(C(O)=O)C1=CC=CC=C1
Color White
Melting Point 117.0°C to 120.0°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula C6H5CH(CH2OH)CO2H
Beilstein 10, 261
Merck Index 15, 9958
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,practically insoluble in petroleum ether
Formula Weight 166.18
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material DL-Tropic acid
Show More Show Less

GHS H StatementCauses skin irritation.Causes serious eye irritation.May cause respiratory irritation.

GHS Signal Word: Warning