Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Tropic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7026184
Click to view available options
Quantity:
10 g
50 g
552-63-6
C9H10O3
166.18
MFCD00004255
JACRWUWPXAESPB-UHFFFAOYNA-N
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid
10726
CHEBI:30765
3-hydroxy-2-phenylpropanoic acid
OCC(C(O)=O)C1=CC=CC=C1
552-63-6
C9H10O3
166.18
MFCD00004255
JACRWUWPXAESPB-UHFFFAOYNA-N
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid
10726
CHEBI:30765
3-hydroxy-2-phenylpropanoic acid
OCC(C(O)=O)C1=CC=CC=C1

It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.

Solubility
Soluble in water (20 g/L) at 20°C, ethanol, ether, alcohol, and methanol (0.1 g/ml).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 117°C to 119°C
Quantity 10 g
Beilstein 2209199
Merck Index 14,9779
Solubility Information Soluble in water (20g/L) at 20°C,ethanol,ether,alcohol,and methanol (0.1g/ml).
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material DL-Tropic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-020-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit