Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ DL-Tryptophan methyl ester hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
5619-09-0
C12H15ClN2O2
254.714
MFCD00066133
XNFNGGQRDXFYMM-UHFFFAOYSA-N
dl-tryptophan methyl ester hydrochloride,methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride,h-dl-trp-ome.hcl,dl-tryptophan methyl ester hcl,h-d-trp-ome hcl,methyl 2-amino-3-indol-3-ylpropanoate, chloride,h-dl-trp-ome hcl,h-dl-trp-ome-hcl,pubchem10907,acmc-209oxn
2827264
methyl 2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
COC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
5619-09-0
C12H15ClN2O2
254.714
MFCD00066133
XNFNGGQRDXFYMM-UHFFFAOYSA-N
dl-tryptophan methyl ester hydrochloride,methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride,h-dl-trp-ome.hcl,dl-tryptophan methyl ester hcl,h-d-trp-ome hcl,methyl 2-amino-3-indol-3-ylpropanoate, chloride,h-dl-trp-ome hcl,h-dl-trp-ome-hcl,pubchem10907,acmc-209oxn
2827264
methyl 2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
COC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
Formula Weight 268.74
Physical Form Solid
Percent Purity 97%
Color White
Melting Point 221°C to 222°C
Quantity 25g
Chemical Name or Material DL-Tryptophan methyl ester hydrochloride