Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLAT Mouse anti-Human, Mouse, Clone:, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524764
Description
DLAT encodes component E2 of the multi-enzyme pyruvate dehydrogenase complex (PDC). PDC resides in the inner mitochondrial membrane and catalyzes the conversion of pyruvate to acetyl coenzyme A. The protein product of this gene, dihydrolipoamide acetyltransferase, accepts acetyl groups formed by the oxidative decarboxylation of pyruvate and transfers them to coenzyme A. Dihydrolipoamide acetyltransferase is the antigen for antimitochondrial antibodies. These autoantibodies are present in nearly 95% of patients with the autoimmune liver disease primary biliary cirrhosis (PBC). In PBC, activated T lymphocytes attack and destroy epithelial cells in the bile duct where this protein is abnormally distributed and overexpressed. PBC enventually leads to cirrhosis and liver failure.Specifications
DLAT | |
Monoclonal | |
0.1M tris glycine with 150mM NaCl, 50% glycerol and 0.02% sodium azide | |
P10515, Q8BMF4 | |
DLAT | |
Human DLAT recombit protein | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
DLAT | |
70 kDa mitochondrial autoantigen of primary biliary cirrhosis;anti DLAT; Dihydrolipoamide acetyltransferase component of pyruvate dehydrogenase complex; Dihydrolipoamide;Dihydrolipoamide S Acetyltransferase;Dihydrolipoamide S-acetyltransferase (E2 component of pyruvate dehydrogenase complex);Dihydrolipoamide S-Acetyltransferase;Dihydrolipoyllysine-residue acetyltransferase component of pyruvate dehydrogenase complex;dihydrolipoyllysine-residue acetyltransferase component of pyruvate dehydrogenase complex mitochondrial;DLAT;DLAT;DLTA;E2;E2 component of pyruvate dehydrogenase complex;EC 2.3.1.12;M2 antigen complex 70 kDa subunit;M2 Antigen Complex 70kD Subunit;mitochondrial;ODP2_HUMAN;PBC;PDC E2;PDC-E2;PDCE2;Pyruvate dehydrogenase complex component E2;Pyruvate dehydrogenase complex E2 subunit;S acetyltransferase component of pyruvate dehydrogenase complex. | |
Mouse | |
Purified | |
RUO | |
1737, 235339 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction