Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLD Mouse anti-Human, Clone: OTI4E11, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503390
Description
This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.Specifications
DLD | |
Monoclonal | |
0.57 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P09622 | |
DLDD, DLDH, E3, GCSL, LAD, PHE3 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
1738 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4E11 | |
Unconjugated | |
DLD | |
Liquid | |
DLD | |
Full length human recombit protein of human DLD produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title