Learn More
DLK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156801AP
Catalog No. 50-173-0399
Add to cart
Description
Description
Regulates adipogenesis.
Specifications
Specifications
DLK2
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZUK3, Q6UY11, Q8K1E3
DLK 2, DLK2, EGF like protein 9, EGFL9, Protein delta homolog 2
Rabbit
IgG
150 μL
-20°C
Primary
106565, 316232, 65989
Immunoprecipitation, Western Blot
Unconjugated
DLK2
Liquid
Dlk2
DLK2 Fusion Protein Ag8170
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat