Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLL1 Antibody (D1L165-6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280048F
This item is not returnable.
View return policy
Description
DLL1 Monoclonal antibody specifically detects DLL1 in Human samples. It is validated for ELISASpecifications
DLL1 | |
Monoclonal | |
FITC | |
Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
Recombinant human DLL1 | |
0.1 mL | |
Neuroscience, Signal Transduction, Stem Cell Signaling Pathway | |
28514 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
D1L165-6 | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction