Learn More
DLL1 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0247- 2X38, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212266AF350
Catalog No. NP212266A30
Add to cart
Description
Description
DLL1 Monoclonal antibody specifically detects DLL1 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
DLL1
MM0247- 2X38
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
DLL1
Human DLL1
0.1 mL
Primary
28514