Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLL1 Antibody (MM0247- 2X38), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212266AF532
Description
DLL1 Monoclonal specifically detects DLL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DLL1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
DLL1 | |
Human recombinant DLL1 | |
Primary | |
Human | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0247- 2X38 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
Mouse | |
0.1 mL | |
28514 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction