Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group Inc 202301AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-2782


Add to cart

Description

Description

The antibody is specific to the protein DLL1.

DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may play a role in cell-to-cell communication.
Specifications

Specifications

DLL1
Polyclonal
Unconjugated
DLL1
Delta, delta like 1 (Drosophila), Delta like protein 1, DELTA1, DL1, DLL1, Drosophila Delta homolog 1, H Delta 1
Rabbit
Antigen Affinity Chromatography
RUO
13388, 28514
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00548, Q61483
DLL1
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit