Learn More
Aviva Systems Biology Corp DLL3 Antibody - N-terminal region (ARP47292_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DLL3. Alternate names: SCDO1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59510