Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLL4 Mouse anti-Human, PE/Atto594, Clone: MHD4-46, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262229PEATT594

Catalog No. N262229P594


Add to cart

Description

Description

DLL4 Monoclonal antibody specifically detects DLL4 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

DLL4
MHD4-46
Flow Cytometry
Conjugated, Purified
DLL4
recombinant soluble human DLL4
Protein A purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
The antibody GF-01 reacts with GFAP, the principal marker of astroglial cells in the central nervous system, which is specifically expressed in satellite cells in peripheral ganglia and in non myelinating Schwann cells in peripheral nerves. The GFAP protein runs on gels at ∽55 kDa protein, usually associated with lower Mw bands which are thought to be proteolytic fragments and alternate transcripts from the single gene.
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4
Mouse
IgG1 Kappa
0.1 mL
Primary
54567.0
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit