DMAB-anabaseine dihydrochloride, Tocris

Partial agonist at α7-containing receptors and antagonist at α4β2 neuronal nicotinic receptors

Manufacturer: R&D Systems 2241/10

Quantity 10mg

 View more versions of this product

Catalog No. 22-411-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C19H21N3·2HCl
    Formula Weight 364.32
    Synonym 4-[(5,6-Dihydro[2,3'-bipyridin]-3(4H)-ylidene)methyl]-N,N-dimethylbenzenamine dihydrochloride
    Chemical Name or Material DMAB-anabaseine dihydrochloride

    Specifications

    Formula Weight 364.32
    Quantity 10mg
    Synonym 4-[(5,6-Dihydro[2,3'-bipyridin]-3(4H)-ylidene)methyl]-N,N-dimethylbenzenamine dihydrochloride
    Assay Percent Range >98%
    Molecular Formula C19H21N3·2HCl
    CAS 154149-38-9
    Recommended Storage Desiccate at -20°C
    Chemical Name or Material DMAB-anabaseine dihydrochloride

    Desiccate at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit