Learn More
Aviva Systems Biology Corp DMAP1 ANTIBODY 100UG/100UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP32917_T100
This is a rabbit polyclonal antibody against DMAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DMAP1. Alternate names: EAF2, SWC4, MEAF2, DNMAP1, DNMTAP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54819