Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMBT1/GP340 Mouse anti-Human, Alexa Fluor™ 594, Clone: 1G4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223576AF594

Catalog No. NP22376A94


Add to cart

Description

Description

DMBT1/GP340 Monoclonal antibody specifically detects DMBT1/GP340 in Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry.
Specifications

Specifications

DMBT1/GP340
1G4
Western Blot, ELISA, Immunohistochemistry
deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, FLJ61058, Glycoprotein 340, gp-340, GP340muclin, Hensin, MGC164738, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 1G4 Reacts with gp-340 and salivary agglutinin
ELISA, Immunohistochemistry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
DMBT1
Native gp-340 purified from bronchoalveolar lavage fluid from patients with pulmonary alveolar proteinosis, adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
1755
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit