Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMBT1/GP340 Antibody (1G4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
$409.50
Specifications
Antigen | DMBT1/GP340 |
---|---|
Clone | 1G4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP22357601
|
Novus Biologicals
NBP2235760.1ML |
0.1 mL |
Each of 1 for $409.50
|
|
|||||
Description
DMBT1/GP340 Monoclonal specifically detects DMBT1/GP340 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
DMBT1/GP340 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, FLJ61058, Glycoprotein 340, gp-340, GP340muclin, Hensin, MGC164738, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein | |
DMBT1 | |
IgG1 κ | |
Protein A or G purified | |
Clone 1G4 Reacts with gp-340 and salivary agglutinin |
1G4 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 | |
Monoclonal | |
Purified | |
Breast Cancer | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
1755 | |
Native gp-340 purified from bronchoalveolar lavage fluid from patients with pulmonary alveolar proteinosis, adsorbed onto aluminum hydroxide gel | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title