Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMBT1/GP340 Mouse anti-Human, Janelia Fluor 549, Clone: 1G4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223576JF549

Catalog No. NBP23576549


Add to cart

Description

Description

DMBT1/GP340 Monoclonal antibody specifically detects DMBT1/GP340 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

DMBT1/GP340
1G4
Western Blot, ELISA, Immunohistochemistry
Purified
DMBT1
Native gp-340 purified from bronchoalveolar lavage fluid from patients with pulmonary alveolar proteinosis, adsorbed onto aluminum hydroxide gel
Protein A or G purified
Breast Cancer
Primary
1755
Human
ELISA, Immunohistochemistry, Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, FLJ61058, Glycoprotein 340, gp-340, GP340muclin, Hensin, MGC164738, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone 1G4 Reacts with gp-340 and salivary agglutinin
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit