Learn More
DMBT1/GP340 Mouse anti-Human, Janelia Fluor 549, Clone: 5D7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP105148JF549
Catalog No. NBP15148549
Add to cart
Description
Description
DMBT1/GP340 Monoclonal antibody specifically detects DMBT1/GP340 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
DMBT1/GP340
5D7
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
DMBT1
Native gp-340 purified from bronchioaveolar lavage from proteinosis patients
Protein A or G purified
Breast Cancer
Primary
1755
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, FLJ61058, Glycoprotein 340, gp-340, GP340muclin, Hensin, MGC164738, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Specific for gp-340, but reacts with a carbohydrate epitope and cross-reacts with saliva MUC7 in immunohistochemistry.