Learn More
BioworldSupplier Diversity Partner DMEM, High Gluc w/L-glut, Pyridoxine HCL and HEPES Buffer w/o Sodium Pyruvate and Phenol Red, 50 L DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306110043
DMEM (High Glucose with L-glutamine); With Pyridoxine Hydrochloride and HEPES Buffer, No Sodium Pyruvate and Phenol Red; Powder. Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME), containing four-fold concentrations of the amino acids and vitamins found in BME, as well as additional supplementary components. The original formulation of DMEM contained 1000mg/L of glucose, but since has been raised in some modifications to 4500mg/L, giving the terms of Low Glucose or High Glucose. Several other modifications over the years, in addition to the alternative glucose levels, have broadened the spectrum of cell types supported by DMEM.
Catalog No. 50-254-026