Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMF, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
624-49-7
C6H8O4
144.126
LDCRTTXIJACKKU-ONEGZZNKSA-N
dimethyl fumarate,tecfidera,e-dimethyl fumarate,fumaderm,dimethyl e-but-2-enedioate,fumaric acid, dimethyl ester,boletic acid dimethyl ester,fumaric acid dimethyl ester,methyl fumarate,dimethyl trans-ethylenedicarboxylate
637568
CHEBI:76004
dimethyl (E)-but-2-enedioate
COC(=O)C=CC(=O)OC
This item is not returnable. View return policy
624-49-7
C6H8O4
144.126
LDCRTTXIJACKKU-ONEGZZNKSA-N
dimethyl fumarate,tecfidera,e-dimethyl fumarate,fumaderm,dimethyl e-but-2-enedioate,fumaric acid, dimethyl ester,boletic acid dimethyl ester,fumaric acid dimethyl ester,methyl fumarate,dimethyl trans-ethylenedicarboxylate
637568
CHEBI:76004
dimethyl (E)-but-2-enedioate
COC(=O)C=CC(=O)OC

Nrf2 pathway activator; neuroprotective

Nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator; induces upregulation of antioxidant gene expression. Displays cytoprotective effects in human spinal cord astrocytes, oligodendrocyte precursor cells and hippocampal neurons following hydrogen peroxide challenge. Exerts neuroprotective effects in vivo in experimental autoimmune encephalomyelitis. Primary metabolite ( MMF, Cat. No. 4511) available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 144.13
Percent Purity >99%
Chemical Name or Material DMF

Recommended Storage : Store at Room Temperature