Learn More
Tocris Bioscience™ DMH2 Available on GSA/VA Contract for Federal Government customers only.
Type I BMP receptor inhibitor
$209.00 - $869.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
558050 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5580/50
Each for $869.50
Add to cart
558010 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5580/10
Each for $209.00
Add to cart
Description
Description
Type I BMP receptor inhibitor (Ki values are <1, 5.4 and 43 nM for ALK6, ALK3 and ALK2, respectively). Exhibits >30-fold selectivity over ALK4, ALK5, BMPR2, AMPK and VEGFR2. Also binds to TGFBR2 (Ki = 85 nM). Decreases growth and induces cell death of H1299 non-small cell lung carcinoma cells in vitro. Enhances liver regeneration in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.