Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMKN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597666
Description
DMKN Polyclonal Antibody for Western Blot, ELISA
The function of this protein is still unknown. The human ZD52F10 gene is up-regulated in colon cancer metastasis. It encode the protein (dermokine), which is a new stratified epithelium-secreted protein.
Specifications
DMKN | |
Polyclonal | |
Unconjugated | |
Dmkn | |
1110014F24Rik; AW561900; C130074A08; cI-36; Dermokine; Dmkn; Epidermis-specific secreted protein SK30/SK89; RGD1561521; SK30; SK89; UNQ729; UNQ729/PRO1411; ZD52F10 | |
Rabbit | |
Protein G | |
RUO | |
73712, 93099 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q6E0U4, Q6P253 | |
Dmkn | |
Recombinant Human Dermokine protein (1-137AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction