Learn More
Cole-Parmer Amprobe 33XR DMM, Prof with Temp & Capacitance  DFS Item
Manufacturer:  Cole-Parmer 2000547
This Amprobe features a frequency to 40 MHz; temperature to 1832 F, and capacitance to 4000 F. It also has the following features: 1,000 V/750 V dc/ac, 10 Amps ac/dc, fully safety fused; resistance to 4 M; continuity beeper, and input warning tone when test leads are in wrong jack.
Catalog No. 50-630-886