Learn More
Cole-Parmer Amprobe 38XR-A DMM T-RMS W/OPTIC PC INTERFAC  DFS Item
Manufacturer:  Cole-Parmer 2000551
Amprobe 38XR-A, a true-rms digital multimeter with optical PC interface for engineers designing and troubleshooting industrial automation and control systems.
Catalog No. 50-630-890