Learn More
DMP-1 Mouse anti-Human, Alexa Fluor™ 350, Clone: 35K29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221973AF350
Catalog No. NP221973A30
Add to cart
Description
Description
DMP-1 Monoclonal antibody specifically detects DMP-1 in Human samples. It is validated for ELISA.
Specifications
Specifications
DMP-1
35K29
ELISA
ARHP, ARHR, dentin matrix acidic phosphoprotein, dentin matrix acidic phosphoprotein 1, Dentin matrix protein 1, DMP-1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
DMP1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant DMP-1. Mouse anti human DMP-1 NP_001073380.
0.1 mL
Primary
1758