Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMP 543, Tocris Bioscience™

Catalog Number p-4783915
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
160588-45-4
C26H18F2N2O
412.44
MUJBUUDUXGDXLW-UHFFFAOYSA-N
unii-dyd4418j7v, 10,10-bis 2-fluoro-4-pyridinyl methyl-9 10h-anthracenone, 10,10-bis 2-fluoropyridin-4-yl methyl anthracen-9 10h-one, d0w7ir, 10,10-bis 2-fluoropyridin-4-yl methyl anthracen-9-one, 10,10-bis 2-fluoro-4-pyridyl methyl anthrone, 10,10-bis 2-fluoro-4-pyridinylmethyl-9 10h-anthracenone, 9 10h-anthracenone, 10,10-bis 2-fluoro-4-pyridinyl methyl
9887884
10,10-bis[(2-fluoropyridin-4-yl)methyl]anthracen-9-one
C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3C2(CC4=CC(=NC=C4)F)CC5=CC(=NC=C5)F
This item is not returnable. View return policy
160588-45-4
C26H18F2N2O
412.44
MUJBUUDUXGDXLW-UHFFFAOYSA-N
unii-dyd4418j7v, 10,10-bis 2-fluoro-4-pyridinyl methyl-9 10h-anthracenone, 10,10-bis 2-fluoropyridin-4-yl methyl anthracen-9 10h-one, d0w7ir, 10,10-bis 2-fluoropyridin-4-yl methyl anthracen-9-one, 10,10-bis 2-fluoro-4-pyridyl methyl anthrone, 10,10-bis 2-fluoro-4-pyridinylmethyl-9 10h-anthracenone, 9 10h-anthracenone, 10,10-bis 2-fluoro-4-pyridinyl methyl
9887884
10,10-bis[(2-fluoropyridin-4-yl)methyl]anthracen-9-one
C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3C2(CC4=CC(=NC=C4)F)CC5=CC(=NC=C5)F

Potent, orally active ACh release enhancer

Acetylcholine release stimulator. Potently enhances K+-stimulated [3H]-ACh release from rat hippocampal slices (EC50 = 700 nM), and increases release of dopamine and glutamate (EC50 values are 0.25 and 0.22 μM, respectively). Orally active in vivo; increases ACh levels in rats (with a minimum effective dose of 1 mg/kg) and exerts a long duration of action. More potent than linopirdine (Cat. No. 1999) both in vitro and in vivo. Also K+ channel blocker.
Quantity 10 mg
Formula Weight 412.44
Percent Purity >99%
Chemical Name or Material DMP 543
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit