Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMP 543, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Acetylcholine release stimulator. Potently enhances K+-stimulated [3H]-ACh release from rat hippocampal slices (EC50 = 700 nM), and increases release of dopamine and glutamate (EC50 values are 0.25 and 0.22 μM, respectively). Orally active in vivo; increases ACh levels in rats (with a minimum effective dose of 1 mg/kg) and exerts a long duration of action. More potent than linopirdine (Cat. No. 1999) both in vitro and in vivo. Also K+ channel blocker.
Specifications
Quantity | 10 mg |
Formula Weight | 412.44 |
Percent Purity | >99% |
Chemical Name or Material | DMP 543 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction