Learn More
DMPO Nitrone Adduct Mouse anti-Mouse, Non-species specific, Biotin, Clone: N1664A, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259306B
Catalog No. NBP259306B
Add to cart
Description
Description
DMPO Nitrone Adduct Monoclonal antibody specifically detects DMPO Nitrone Adduct in Mouse, Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
DMPO Nitrone Adduct
N1664A
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
5,5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated to Ovalbumin
Protein G purified
Store at 4C in the dark.
Monoclonal
Mouse, Non-species specific
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Recognizes DMPO, DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Does not cross react with non-adducted proteins or DNA.