Learn More
DMPO Nitrone Adduct Mouse anti-Mouse, Non-species specific, Clone: N1664A, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259306
Catalog No. NBP259306
Add to cart
Description
Description
DMPO Nitrone Adduct Monoclonal antibody specifically detects DMPO Nitrone Adduct in Mouse, Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
DMPO Nitrone Adduct
N1664A
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Purified
5,5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated to Ovalbumin
Protein G purified
RUO
Primary
Recognizes DMPO, DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Does not cross react with non-adducted proteins or DNA.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Mouse
IgG1
100 ug
Store at -20C.
Monoclonal
Mouse, Non-species specific