Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMRT1 Antibody (OTI1G10), DyLight 680, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270582FR

Catalog No. NBP270582FR


Add to Cart

Description

Description

DMRT1 Monoclonal specifically detects DMRT1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DMRT1
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
DMRT1
Recombinant protein fragment corresponding to amino acids 1-210 of human DMRT1 (NP_068770) produced in E.coli.
0.1 mL
1761.0
Store at 4°C in the dark.
Western Blot
OTI1G10
Western Blot
DM domain expressed in testis 1, DMT1DM domain expressed in testis protein 1, doublesex and mab-3 related transcription factor 1, doublesex- and mab-3-related transcription factor 1
Mouse
Affinity Purified
Primary
Human
IgG2b
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit