Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMRT1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1G10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270582AF405

Catalog No. N270582A405


Add to cart

Description

Description

DMRT1 Monoclonal antibody specifically detects DMRT1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DMRT1
OTI1G10
Western Blot
Affinity Purified, Conjugated
DMRT1
Recombinant protein fragment corresponding to amino acids 1-210 of human DMRT1 (NP_068770) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
DM domain expressed in testis 1, DMT1DM domain expressed in testis protein 1, doublesex and mab-3 related transcription factor 1, doublesex- and mab-3-related transcription factor 1
Mouse
IgG2b
0.1 mL
Primary
1761.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit