Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMRTA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566545
Description
Immunogen sequence: VPTLGFRPPMD YAFSDLMRDR SAAAAAAVHK EPTYGGGLYG PMVNGAPEKQ Highest antigen sequence identity to the following orthologs - mouse 96%, rat 30%.
DMRTA2 may be involved in sexual development.
Specifications
DMRTA2 | |
Polyclonal | |
Unconjugated | |
DMRTA2 | |
DMRT like family A2; Dmrt5; Dmrta2; DMRT-like family A2; doublesex and mab-3 related transcription factor like family A2; doublesex- and mab-3-related transcription factor 5; Doublesex- and mab-3-related transcription factor A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63950 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96SC8 | |
DMRTA2 | |
Recombinant Human DMRTA2. Recombinant protein control fragment (Product #RP-107207). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction