Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMTF1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen DMTF1
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125570
View Documents
Novus Biologicals
NBP31033425UL
25 μg
Each for $106.19
Add to cart
 
NB125569
View Documents
Novus Biologicals
NBP310334100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

DMTF1 Polyclonal antibody specifically detects DMTF1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

DMTF1
Western Blot, Immunohistochemistry
Purified
RUO
Human
cyclin D binding myb-like transcription factor 1, cyclin D-binding Myb-like protein 1, cyclin-D-binding Myb-like transcription factor 1, Cyclin-D-interacting Myb-like protein 1, DMP1FLJ76054, FLJ25188, FLJ41265, hDMP1DMTF, hDMTF1
The immunogen is a synthetic peptide directed towards the C terminal region of human DMTF1 (EAW76962). Peptide sequence SFNDAHVSKFSDQNSTELMNSVMVRTEEEISDTDLKQEESPSDLASAYVT
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Breast Cancer
PBS buffer, 2% sucrose
9988
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit