Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DMXAA

mSTING agonist; induces antitumor immunological responses

$148.90 - $627.00

Specifications

Chemical Name or Material 5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic acid
Molecular Formula C17H14O4
CAS 117570-53-3
Product Type Target: Cytokine Compounds
Formula Weight 282.29
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
560110 View Documents Tocris Bioscience™
5601/10
10mg Each for $148.90
Add to cart
 
560150 View Documents Tocris Bioscience™
5601/50
50mg Each for $627.00
Add to cart
 
Description

Description

mSTING agonist; selective for mouse STING over human STING. Induces IFN-β and cytokine production from bone marrow-derived dendritic cells. Causes tumor regression following intratumoral administration in mouse tumor models. Induces an adaptive immune response to reduce systemic tumor growth and provides immunological memory against autologous tumor re-challenge. Also antiviral.
Specifications

Specifications

5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic acid
117570-53-3
282.29
C17H14O4
Target: Cytokine Compounds
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit