Learn More
Thermo Scientific™ DNA AWAY™ Surface Decontaminant
Eliminate unwanted DNA and DNase from glassware and plasticware without affecting subsequent DNA samples. This surface decontaminant degrades DNA more quickly and effectively than autoclaving.
Manufacturer:  Thermo Scientific™ 7008
 View more versions of this product
Catalog No. 14-754-42
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Eliminate unwanted DNA and DNase from glassware and plastic ware without affecting subsequent DNA samples. Thermo Scientific™ DNA AWAY™ Surface Decontaminant degrades DNA more quickly and effectively than autoclaving.
  • Suitable for gel boxes, pipettes, benchtops, thermal cyclers, or other apparatus
  • Use before performing PCR or working with DNA for a DNA-free work area
  • Simply apply the ready-to-use formula to the surface to decontaminate, then wipe dry or rinse clean
Specifications
Specifications
Decontaminant Wipes
25/Pk.