Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA, Fish Sperm, Sodium Salt, Affymetrix/USB™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
68938-01-2
C17H15BrFNO3
380.213
MFCD00284534
GXZCYECDYOBPGH-OAHLLOKOSA-N
73904662
(2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
C1=CC(=CC(=C1)Br)CC(C(=O)O)NC(=O)CC2=CC=C(C=C2)F
68938-01-2
C17H15BrFNO3
380.213
MFCD00284534
GXZCYECDYOBPGH-OAHLLOKOSA-N
73904662
(2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
C1=CC(=CC(=C1)Br)CC(C(=O)O)NC(=O)CC2=CC=C(C=C2)F
Physical Form Powder
Packaging Plastic bottle
Quantity 100g
Chemical Name or Material DNA