Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase I Mouse anti-Human, Mouse, Rat, Bovine, Clone: 1A9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30724810UG
Description
DNA Ligase I Monoclonal specifically detects DNA Ligase I in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
DNA Ligase I | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, ELISA, Immunoprecipitation | |
DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 | |
Full-length native cow LDNA Ligase I protein | |
100 μg | |
Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
3978 | |
Store at 4°C. | |
IgG1 κ |
Western Blot, ELISA, Immunoprecipitation | |
1A9 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title