Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Ligase I Mouse anti-Human, Mouse, Rat, Bovine, Clone: 1A9, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen DNA Ligase I
Clone 1A9
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA, Immunoprecipitation
Applications Western Blot, ELISA, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121969
View Documents
Novus Biologicals
NBP30724810UG
100 μg
Each for $352.89
Add to cart
 
NB121970
View Documents
Novus Biologicals
NBP30724820UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

DNA Ligase I Monoclonal antibody specifically detects DNA Ligase I in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications

Specifications

DNA Ligase I
0.2 mg/ml
Western Blot, ELISA, Immunoprecipitation
Purified
RUO
Human, Mouse, Rat, Bovine
DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1
Full-length native cow LDNA Ligase I protein
Monoclonal
Protein A or G purified
1A9
Western Blot 1-2 ug/ml, ELISA, Immunoprecipitation
Unconjugated
Mouse
Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors
10 mM PBS with 0.05% BSA
3978
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit