Learn More
Eppendorf™ DNA LoBind Tubes
DNA LoBind Tubes maximize sample recovery of nucleic acids by reducing sample-to-surface binding. Free of surface coating for less sample interference and precise lid sealing for minimal evaporation.
Manufacturer:  Eppendorf™ 2231000347
 View more versions of this product
Catalog No. 05-412-698
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Features:
  • Eppendorf LoBind material for maximum sample recovery and improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Precise lid sealing for minimal evaporation rates in tube format
  • Rated up to 30,000 x g (25,000 x g for 2.0mL tubes) centrifugation speed for molecular biology applications
Features: 
  • Eppendorf LoBind material for maximum sample recovery and improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Precise lid sealing for minimal evaporation rates in tube format
  • Rated up to 30,000 x g (25,000 x g for 2.0mL tubes) centrifugation speed for molecular biology applications
Specifications
Specifications
Snap top
Colorless
Centrifuge
LoBind
5.0mL
Preparation and storage of DNA and RNA samples; Forensic trace analysis; Dilution series in qPCR; Next-generation sequencing; Genomic or oligonucleotide libraries
250 (5 bags x 50)
PCR clean