Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ DNA-PKcs Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments.

Specifications

Antigen DNA-PKcs Ab-2
Clone 25-4
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS370P
View Documents
Thermo Scientific™
MS370P
200μg/mL 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS370P0
View Documents
Thermo Scientific™
MS370P0
200μg/mL 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS370P1
View Documents
Thermo Scientific™
MS370P1
200μg/mL 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS370P1ABX
View Documents
Thermo Scientific™
MS370P1ABX
1mg/mL 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS370PABX
View Documents
Thermo Scientific™
MS370PABX
1mg/mL 200μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ DNA-PKcs Ab-2, Mouse Monoclonal Antibody.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 25-4

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: aa 3198-4127

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift (Not optimum)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

DNA-PKcs Ab-2
Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Murine
DNA Repair
Human DNA-dependent Protein Kinase purified from HeLa cells
25-4
Monoclonal
Concentrated
RUO
Primary
IgG2a
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit