Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase delta, catalytic subunit Mouse anti-Human, DyLight 488, Clone: 6196, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243556G

Catalog No. NBP243556G


Add to cart

Description

Description

DNA Polymerase delta, catalytic subunit Monoclonal antibody specifically detects DNA Polymerase delta, catalytic subunit in Human samples. It is validated for Western Blot.
Specifications

Specifications

DNA Polymerase delta, catalytic subunit
6196
Western Blot
Affinity Purified
POLD1
Recombinant protein encoding amino acids 1-140 of the large subunit of human DNA polymerase d.
Immunogen affinity purified
Breast Cancer, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers
Primary
5424
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
CDC2, CDC2 homolog, DNA polymerase delta catalytic subunit, DNA polymerase subunit delta p125, DNA-directed polymerase delta 1, EC 2.7.7.7, POLD, polymerase (DNA directed), delta 1, catalytic subunit (125kD), polymerase (DNA directed), delta 1, catalytic subunit 125kDa
Mouse
IgG
100 ul
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit