Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase epsilon catalytic subunit A, Mouse anti-Human,Mouse,Mustelid,Primate, Alexa Fluor 647, Clone: 3C5.1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB100115AF6470.1ML

Catalog No. NB003402


Add to cart

Description

Description

DNA Polymerase epsilon catalytic subunit A Monoclonal specifically detects DNA Polymerase epsilon catalytic subunit A in Human, Mouse, Mustelid, Primate, Drosophila (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

DNA Polymerase epsilon catalytic subunit A
3C5.1
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
POLE
Human DNA Polymerase epsilon purified from HeLa cells.
Protein G purified
RUO
Store at 4°C in the dark.
Monoclonal
This is specific to the human DNA pol epsilon and does not recognize DNA pol alpha, beta or delta.
Western Blot, Immunofluorescence
Alexa Fluor 647
50mM Sodium Borate
DKFZp434F222, DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A, EC 2.7.7, EC 2.7.7.7, POLE1FLJ21434, polymerase (DNA directed), epsilon
Mouse
IgG2a
0.1 mL
Chromatin Research, DNA Polymerases, DNA Repair
Primary
5426
Human, Mouse, Mustelid, Primate, Drosophila (Negative)
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit