Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase epsilon subunit 3 Mouse anti-Human, Clone: PCRP-POLE3-2F10, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen DNA Polymerase epsilon subunit 3
Clone PCRP-POLE3-2F10
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122706
View Documents
Novus Biologicals
NBP30761620UG
20 μg
Each for $138.37
Add to cart
 
NB122705
View Documents
Novus Biologicals
NBP30761610UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

DNA Polymerase epsilon subunit 3 Monoclonal antibody specifically detects DNA Polymerase epsilon subunit 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

DNA Polymerase epsilon subunit 3
0.2 mg/ml
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
Arsenic-transactivated protein, asTP, CHARAC17arsenic transactivated protein, CHRAC-17, CHRAC17Ybl1, Chromatin accessibility complex 17 kDa protein, DNA polymerase epsilon p17 subunit, DNA polymerase epsilon subunit 3, DNA polymerase epsilon subunit p17, DNA polymerase II subunit 3, EC 2.7.7.7, histone fold protein CHRAC17, HuCHRAC17, p17, polymerase (DNA directed), epsilon 3 (p17 subunit), YBL1
Recombinant full-length human DNA Polymerase epsilon subunit 3 protein
Monoclonal
Protein A or G purified
PCRP-POLE3-2F10
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array
Unconjugated
Mouse
Chromatin Research, DNA Polymerases, DNA Repair
10 mM PBS with 0.05% BSA
54107
IgG1
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit