Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DNA Polymerase I Large (Klenow Fragment) Kit

Specifications

Content And Storage Store at -20°C
Format Liquid
Packaging Plastic bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ65056ZEX
View Documents
Thermo Scientific™
J65056ZEX
150 units Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
AAJ65056EXU
View Documents
Thermo Scientific™
J65056EXU
500 units Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Store at -20°C
Liquid
Plastic bottle
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit