Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA polymerase mu Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | DNA polymerase mu |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
DNA polymerase mu Polyclonal specifically detects DNA polymerase mu in Human samples. It is validated for Western Blot.Specifications
DNA polymerase mu | |
Polyclonal | |
Rabbit | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
DNA polymerase mu, DNA-directed DNA/RNA polymerase mu, EC 2.7.7.7, FLJ35482, Pol iota, Pol Mu, polmu, polymerase (DNA directed), mu, polymerase (DNA-directed), mu, Tdt-N, Terminal transferase | |
POLM | |
IgG | |
55 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9NP87 | |
27434 | |
Synthetic peptides corresponding to POLM(polymerase (DNA directed), mu) The peptide sequence was selected from the middle region of POLM (NP_037416). Peptide sequence LRRFSRKEKGLWLNSHGLFDPEQKTFFQAASEEDIFRHLGLEYLPPEQRN. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title