Learn More
DNA/RNA Damage Mouse anti-Non-species specific, Alexa Fluor 488, Clone: 15A3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11096878AF488
Catalog No. NBT96878488
Add to cart
Description
Description
DNA/RNA Damage Monoclonal antibody specifically detects DNA/RNA Damage in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
DNA/RNA Damage
15A3
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
8-hydroxy-guanosine-BSA and-casein conjugates
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2b
0.1 ml
Primary
Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine).