Learn More
Puritan™ Collection Tube with HydraFlock™ Flocked Swab
Used for collection and storage of samples. Puritan™ DNA/RNA Shield™ Collection Tube with HydraFlock™ Flocked Swab facilitates nucleic acid stability at ambient temperatures and provides microbial inactivation.
Manufacturer:  Puritan™ VL 1201 DSH
Catalog No. 14-960-103
Add to cart
Description
Description
  • Nucleic acid stability at ambient temperatures
  • Total microbial inactivation
  • Compatible with various sample types
HydraFlock Swab
  • Increased surface area on tip
  • Patented multilength fibers
  • Tapered polystyrene handle with breakpoint
Specifications
Specifications
Swab and collection vial
Specimen collection
Multilength fiber
1mL
Self centering cap