Learn More
Enzo Life SciencesSupplier Diversity Partner DNA topoisomerase II alpha/beta monoclonal antibody (AK5) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAMCC215E
Clone: AK5. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human DNA-topoisomerase IIα (N-terminal fragment). UniProt: P11388 (α isozyme), Q02880 (β isozyme). Species Reactivity: Human, Mouse, Rat. Applications: WB. Recommended dilutions: Western Blot (5-10µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-666-455